Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614537 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C10orf11 - chromosome 10 open reading frame 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305581.1 | Intron | NP_001292510.1 | ||||
NM_032024.4 | Intron | NP_114413.1 | ||||
XM_011540256.2 | Intron | XP_011538558.1 | ||||
XM_011540257.2 | Intron | XP_011538559.1 | ||||
XM_017016759.1 | Intron | XP_016872248.1 | ||||
XM_017016760.1 | Intron | XP_016872249.1 | ||||
XM_017016761.1 | Intron | XP_016872250.1 | ||||
XM_017016762.1 | Intron | XP_016872251.1 |