Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602446 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPC5 - glypican 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004466.5 | Intron | NP_004457.1 | ||||
XM_011521054.2 | Intron | XP_011519356.1 | ||||
XM_011521055.2 | Intron | XP_011519357.1 | ||||
XM_011521056.2 | Intron | XP_011519358.1 | ||||
XM_011521057.2 | Intron | XP_011519359.1 | ||||
XM_011521058.2 | Intron | XP_011519360.1 | ||||
XM_011521059.2 | Intron | XP_011519361.1 | ||||
XM_011521060.2 | Intron | XP_011519362.1 | ||||
XM_017020435.1 | Intron | XP_016875924.1 | ||||
XM_017020436.1 | Intron | XP_016875925.1 | ||||
XM_017020437.1 | Intron | XP_016875926.1 |