Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608024 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf201 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C6orf201 - chromosome 6 open reading frame 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085401.2 | 1244 | Intron | NP_001078870.1 | |||
XM_005249112.3 | 1244 | Intron | XP_005249169.1 | |||
XM_011514613.2 | 1244 | Intron | XP_011512915.1 | |||
XM_017010861.1 | 1244 | Intron | XP_016866350.1 |
ECI2 - enoyl-CoA delta isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166010.1 | 1244 | UTR 3 | NP_001159482.1 | |||
NM_006117.2 | 1244 | UTR 3 | NP_006108.2 | |||
NM_206836.2 | 1244 | UTR 3 | NP_996667.2 | |||
XM_006714957.1 | 1244 | UTR 3 | XP_006715020.1 | |||
XM_017010170.1 | 1244 | UTR 3 | XP_016865659.1 |
Set Membership: |
HapMap |