Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613774 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAMSAP1 - calmodulin regulated spectrin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015447.3 | 6811 | UTR 3 | NP_056262.3 | |||
XM_005263396.3 | 6811 | UTR 3 | XP_005263453.1 | |||
XM_005263397.1 | 6811 | UTR 3 | XP_005263454.1 | |||
XM_011518272.1 | 6811 | UTR 3 | XP_011516574.1 | |||
XM_011518273.2 | 6811 | UTR 3 | XP_011516575.1 | |||
XM_011518274.1 | 6811 | UTR 3 | XP_011516576.1 | |||
XM_017014301.1 | 6811 | UTR 3 | XP_016869790.1 | |||
XM_017014302.1 | 6811 | UTR 3 | XP_016869791.1 |