Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616236 MIM: 611865 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHADL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHADL - chondroadherin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138481.1 | Intron | NP_612490.1 | ||||
XM_005261370.3 | Intron | XP_005261427.1 | ||||
XM_011529932.2 | Intron | XP_011528234.1 | ||||
XM_011529933.2 | Intron | XP_011528235.1 | ||||
XM_011529934.2 | Intron | XP_011528236.1 |
L3MBTL2 - L3MBTL2 polycomb repressive complex 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031488.4 | Intron | NP_113676.2 | ||||
XM_011530420.2 | Intron | XP_011528722.1 | ||||
XM_011530421.2 | Intron | XP_011528723.1 | ||||
XM_017028976.1 | Intron | XP_016884465.1 | ||||
XM_017028977.1 | Intron | XP_016884466.1 | ||||
XM_017028978.1 | Intron | XP_016884467.1 | ||||
XM_017028979.1 | Intron | XP_016884468.1 | ||||
XM_017028980.1 | Intron | XP_016884469.1 |
Set Membership: |
HapMap |