Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609818 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP10 - ubiquitin specific peptidase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272075.1 | Intron | NP_001259004.1 | ||||
NM_005153.2 | Intron | NP_005144.2 | ||||
XM_006721332.1 | Intron | XP_006721395.1 | ||||
XM_011523440.1 | Intron | XP_011521742.1 | ||||
XM_011523441.1 | Intron | XP_011521743.1 | ||||
XM_011523443.1 | Intron | XP_011521745.1 | ||||
XM_017023863.1 | Intron | XP_016879352.1 | ||||
XM_017023864.1 | Intron | XP_016879353.1 | ||||
XM_017023865.1 | Intron | XP_016879354.1 | ||||
XM_017023866.1 | Intron | XP_016879355.1 | ||||
XM_017023867.1 | Intron | XP_016879356.1 | ||||
XM_017023868.1 | Intron | XP_016879357.1 | ||||
XM_017023869.1 | Intron | XP_016879358.1 |