Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300642 MIM: 300723 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRPX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SRPX2 - sushi repeat containing protein, X-linked 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL4 - synaptotagmin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129896.2 | 5407 | Intron | NP_001123368.1 | |||
NM_001174068.1 | 5407 | Intron | NP_001167539.1 | |||
NM_080737.2 | 5407 | Intron | NP_542775.2 | |||
XM_005262229.2 | 5407 | UTR 3 | XP_005262286.1 | |||
XM_011531068.2 | 5407 | UTR 3 | XP_011529370.1 | |||
XM_017029966.1 | 5407 | UTR 3 | XP_016885455.1 | |||
XM_017029967.1 | 5407 | UTR 3 | XP_016885456.1 | |||
XM_017029968.1 | 5407 | UTR 3 | XP_016885457.1 | |||
XM_017029969.1 | 5407 | UTR 3 | XP_016885458.1 | |||
XM_017029970.1 | 5407 | UTR 3 | XP_016885459.1 | |||
XM_017029971.1 | 5407 | UTR 3 | XP_016885460.1 |