Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQSEC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IQSEC3 - IQ motif and Sec7 domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170738.1 | Intron | NP_001164209.1 | ||||
NM_015232.1 | Intron | NP_056047.1 | ||||
XM_011520958.2 | Intron | XP_011519260.1 | ||||
XM_011520959.2 | Intron | XP_011519261.1 | ||||
XM_011520960.1 | Intron | XP_011519262.1 | ||||
XM_011520961.1 | Intron | XP_011519263.1 | ||||
XM_017019310.1 | Intron | XP_016874799.1 | ||||
XM_017019311.1 | Intron | XP_016874800.1 |