Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104180 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GANC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GANC - glucosidase alpha, neutral C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301409.1 | 87 | UTR 5 | NP_001288338.1 | |||
NM_001301410.1 | 87 | Intron | NP_001288339.1 | |||
NM_198141.2 | 87 | Intron | NP_937784.2 |
TMEM87A - transmembrane protein 87A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110503.2 | 87 | Intron | NP_001103973.1 | |||
NM_001286487.1 | 87 | Intron | NP_001273416.1 | |||
NM_015497.4 | 87 | Intron | NP_056312.2 | |||
XM_005254287.1 | 87 | Intron | XP_005254344.1 | |||
XM_006720468.2 | 87 | Intron | XP_006720531.1 | |||
XM_011521443.2 | 87 | Intron | XP_011519745.1 | |||
XM_011521444.2 | 87 | Intron | XP_011519746.1 | |||
XM_017022067.1 | 87 | Intron | XP_016877556.1 |
Set Membership: |
HapMap |