Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613880 MIM: 607036 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAHD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BAHD1 - bromo adjacent homology domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301132.1 | 43 | UTR 5 | NP_001288061.1 | |||
NM_014952.4 | 43 | Intron | NP_055767.3 | |||
XM_011521366.2 | 43 | Intron | XP_011519668.2 | |||
XM_011521367.2 | 43 | Intron | XP_011519669.2 | |||
XM_011521368.2 | 43 | Intron | XP_011519670.2 | |||
XM_011521369.2 | 43 | UTR 5 | XP_011519671.1 |
IVD - isovaleryl-CoA dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |