Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607089 MIM: 177070 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNDBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNDBP1 - cyclin D1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPB42 - erythrocyte membrane protein band 4.2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000119.2 | Intron | NP_000110.2 | ||||
NM_001114134.1 | Intron | NP_001107606.1 | ||||
XM_005254225.1 | Intron | XP_005254282.1 | ||||
XM_011521349.2 | Intron | XP_011519651.1 | ||||
XM_011521350.2 | Intron | XP_011519652.1 | ||||
XM_011521351.2 | Intron | XP_011519653.1 | ||||
XM_011521352.2 | Intron | XP_011519654.1 | ||||
XM_011521353.2 | Intron | XP_011519655.1 | ||||
XM_011521354.2 | Intron | XP_011519656.1 |
Set Membership: |
HapMap |