Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606740 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEACAM20 - carcinoembryonic antigen related cell adhesion molecule 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102597.2 | 4121 | Intron | NP_001096067.2 | |||
NM_001102598.2 | 4121 | Intron | NP_001096068.2 | |||
NM_001102599.2 | 4121 | Intron | NP_001096069.2 | |||
NM_001102600.2 | 4121 | Intron | NP_001096070.2 | |||
XM_011526429.2 | 4121 | UTR 3 | XP_011524731.1 | |||
XM_011526430.2 | 4121 | UTR 3 | XP_011524732.1 | |||
XM_011526431.2 | 4121 | Intron | XP_011524733.1 | |||
XM_011526432.2 | 4121 | UTR 3 | XP_011524734.1 |
ZNF180 - zinc finger protein 180 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |