Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612982 MIM: 611780 MIM: 143023 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMNTD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LMNTD2 - lamin tail domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC143666 - uncharacterized LOC143666 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR210 - microRNA 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR210HG - MIR210 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHRF1 - PHD and ring finger domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF7 - Ras association domain family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |