Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602934 MIM: 601333 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDF2 - stromal cell derived factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUPT6H - SPT6 homolog, histone chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320755.1 | Intron | NP_001307684.1 | ||||
NM_003170.4 | Intron | NP_003161.2 | ||||
XM_017024978.1 | Intron | XP_016880467.1 | ||||
XM_017024979.1 | Intron | XP_016880468.1 | ||||
XM_017024980.1 | Intron | XP_016880469.1 |
Set Membership: |
HapMap |