Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGTCAGCCCAGGACACATGGGC[C/T]CCAATGTATTTTGTGTAGCTATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300892 MIM: 300341 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00850 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC00850 - long intergenic non-protein coding RNA 850 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEA8 - MAGE family member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166400.1 | Intron | NP_001159872.1 | ||||
NM_001166401.1 | Intron | NP_001159873.1 | ||||
NM_005364.4 | Intron | NP_005355.2 | ||||
XM_017029520.1 | Intron | XP_016885009.1 |
MAGEA8-AS1 - MAGEA8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |