Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180740 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRNP70 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNRNP70 - small nuclear ribonucleoprotein U1 subunit 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301069.1 | Intron | NP_001287998.1 | ||||
NM_003089.5 | Intron | NP_003080.2 | ||||
XM_005259178.1 | Intron | XP_005259235.1 | ||||
XM_011527240.2 | Intron | XP_011525542.2 | ||||
XM_011527241.2 | Intron | XP_011525543.1 |
Set Membership: |
HapMap |