Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607351 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHOBTB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RHOBTB1 - Rho related BTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242359.1 | Intron | NP_001229288.1 | ||||
NM_014836.4 | Intron | NP_055651.1 | ||||
XM_006718084.1 | Intron | XP_006718147.1 | ||||
XM_006718085.1 | Intron | XP_006718148.1 | ||||
XM_011540424.1 | Intron | XP_011538726.1 | ||||
XM_011540425.1 | Intron | XP_011538727.1 | ||||
XM_011540426.1 | Intron | XP_011538728.1 | ||||
XM_011540428.1 | Intron | XP_011538730.1 | ||||
XM_017017000.1 | Intron | XP_016872489.1 | ||||
XM_017017001.1 | Intron | XP_016872490.1 | ||||
XM_017017002.1 | Intron | XP_016872491.1 | ||||
XM_017017003.1 | Intron | XP_016872492.1 | ||||
XM_017017004.1 | Intron | XP_016872493.1 | ||||
XM_017017005.1 | Intron | XP_016872494.1 | ||||
XM_017017006.1 | Intron | XP_016872495.1 | ||||
XM_017017007.1 | Intron | XP_016872496.1 | ||||
XM_017017008.1 | Intron | XP_016872497.1 |
Set Membership: |
HapMap |