Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602714 MIM: 611640 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ADAM12 - ADAM metallopeptidase domain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288973.1 | 7171 | UTR 3 | NP_001275902.1 | |||
NM_001288974.1 | 7171 | Intron | NP_001275903.1 | |||
NM_001288975.1 | 7171 | Intron | NP_001275904.1 | |||
NM_003474.5 | 7171 | UTR 3 | NP_003465.3 | |||
NM_021641.4 | 7171 | Intron | NP_067673.2 | |||
XM_017016705.1 | 7171 | UTR 3 | XP_016872194.1 | |||
XM_017016706.1 | 7171 | UTR 3 | XP_016872195.1 |
FANK1 - fibronectin type III and ankyrin repeat domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |