Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615662 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SERPINB12 - serpin family B member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307928.1 | Intron | NP_001294857.1 | ||||
NM_080474.2 | Intron | NP_536722.1 | ||||
XM_005266778.3 | Intron | XP_005266835.2 | ||||
XM_011526246.1 | Intron | XP_011524548.1 | ||||
XM_011526248.1 | Intron | XP_011524550.1 | ||||
XM_011526249.1 | Intron | XP_011524551.1 | ||||
XM_017026067.1 | Intron | XP_016881556.1 |
Set Membership: |
HapMap |