Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607567 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA1P5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJA1P5 - DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OLFM3 - olfactomedin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288821.1 | Intron | NP_001275750.1 | ||||
NM_001288823.1 | Intron | NP_001275752.1 | ||||
NM_058170.3 | Intron | NP_477518.2 | ||||
XM_017000240.1 | Intron | XP_016855729.1 |
Set Membership: |
HapMap |