Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614694 MIM: 614425 | ||||||||||||||||||||
Literature Links: |
RPRD1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPRD1B - regulation of nuclear pre-mRNA domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021215.3 | 174 | UTR 5 | NP_067038.1 | |||
XM_005260477.3 | 174 | UTR 5 | XP_005260534.1 | |||
XM_005260480.4 | 174 | UTR 5 | XP_005260537.1 | |||
XM_011528954.2 | 174 | UTR 5 | XP_011527256.1 | |||
XM_017027990.1 | 174 | UTR 5 | XP_016883479.1 | |||
XM_017027991.1 | 174 | UTR 5 | XP_016883480.1 |
TTI1 - TELO2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303457.1 | 174 | Intron | NP_001290386.1 | |||
NM_014657.2 | 174 | Intron | NP_055472.1 | |||
XM_011529114.1 | 174 | Intron | XP_011527416.1 | |||
XM_017028148.1 | 174 | Intron | XP_016883637.1 |