Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608606 MIM: 610989 MIM: 614781 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHLHA15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BHLHA15 - basic helix-loop-helix family member a15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMTK2 - lemur tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TECPR1 - tectonin beta-propeller repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015395.2 | 5069 | UTR 3 | NP_056210.1 | |||
XM_005250253.3 | 5069 | UTR 3 | XP_005250310.1 | |||
XM_017011937.1 | 5069 | UTR 3 | XP_016867426.1 | |||
XM_017011938.1 | 5069 | Intron | XP_016867427.1 |