Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR3B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACTR3B - ARP3 actin related protein 3 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040135.2 | Intron | NP_001035225.1 | ||||
NM_020445.5 | Intron | NP_065178.1 | ||||
XM_006716065.2 | Intron | XP_006716128.1 | ||||
XM_006716066.1 | Intron | XP_006716129.1 | ||||
XM_006716067.3 | Intron | XP_006716130.1 | ||||
XM_011516425.2 | Intron | XP_011514727.1 | ||||
XM_011516426.2 | Intron | XP_011514728.1 | ||||
XM_011516427.1 | Intron | XP_011514729.1 | ||||
XM_017012460.1 | Intron | XP_016867949.1 | ||||
XM_017012461.1 | Intron | XP_016867950.1 | ||||
XM_017012462.1 | Intron | XP_016867951.1 | ||||
XM_017012463.1 | Intron | XP_016867952.1 | ||||
XM_017012464.1 | Intron | XP_016867953.1 | ||||
XM_017012465.1 | Intron | XP_016867954.1 |
Set Membership: |
HapMap |