Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608126 | ||||||||||||||||||||
Literature Links: |
WDHD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDHD1 - WD repeat and HMG-box DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008396.2 | 4063 | UTR 3 | NP_001008397.1 | |||
NM_007086.3 | 4063 | UTR 3 | NP_009017.1 | |||
XM_006720012.2 | 4063 | UTR 3 | XP_006720075.1 | |||
XM_011536373.2 | 4063 | UTR 3 | XP_011534675.1 | |||
XM_017020940.1 | 4063 | UTR 3 | XP_016876429.1 | |||
XM_017020941.1 | 4063 | UTR 3 | XP_016876430.1 |