Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600009 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IFI27 - interferon alpha inducible protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130080.2 | Intron | NP_001123552.1 | ||||
NM_001288952.1 | Intron | NP_001275881.1 | ||||
NM_001288954.1 | Intron | NP_001275883.1 | ||||
NM_001288956.1 | Intron | NP_001275885.1 | ||||
NM_001288957.1 | Intron | NP_001275886.1 | ||||
NM_001288958.1 | Intron | NP_001275887.1 | ||||
NM_001288959.1 | Intron | NP_001275888.1 | ||||
NM_001288960.1 | Intron | NP_001275889.1 | ||||
NM_001288995.1 | Intron | NP_001275924.1 | ||||
NM_005532.4 | Intron | NP_005523.3 |