Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCACAGGCCATTCTCCATTTAA[A/T]TTTTAAATTAAAACAAACATATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602778 | ||||||||||||||||||||
Literature Links: |
MIR181A2HG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR181A2HG - MIR181A2 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR6A1 - nuclear receptor subfamily 6 group A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278546.1 | Intron | NP_001265475.1 | ||||
NM_001489.4 | Intron | NP_001480.3 | ||||
NM_033334.3 | Intron | NP_201591.2 | ||||
XM_005251917.4 | Intron | XP_005251974.1 | ||||
XM_005251918.4 | Intron | XP_005251975.1 |