Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610741 | ||||||||||||||||||||
Literature Links: |
TNRC6C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNRC6C - trinucleotide repeat containing 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142640.1 | Intron | NP_001136112.1 | ||||
NM_018996.3 | Intron | NP_061869.2 | ||||
XM_006721996.3 | Intron | XP_006722059.3 | ||||
XM_006721997.3 | Intron | XP_006722060.3 | ||||
XM_006722000.3 | Intron | XP_006722063.3 | ||||
XM_011525088.2 | Intron | XP_011523390.1 | ||||
XM_017024906.1 | Intron | XP_016880395.1 | ||||
XM_017024907.1 | Intron | XP_016880396.1 | ||||
XM_017024908.1 | Intron | XP_016880397.1 |