Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616864 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEXDC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HEXDC - hexosaminidase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173620.2 | Intron | NP_775891.2 | ||||
XM_005256346.3 | Intron | XP_005256403.1 | ||||
XM_005256347.4 | Intron | XP_005256404.1 | ||||
XM_006722280.3 | Intron | XP_006722343.1 | ||||
XM_011523560.2 | Intron | XP_011521862.1 | ||||
XM_011523561.2 | Intron | XP_011521863.1 | ||||
XM_017024484.1 | Intron | XP_016879973.1 | ||||
XM_017024485.1 | Intron | XP_016879974.1 |