Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615794 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FNDC3A - fibronectin type III domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079673.1 | Intron | NP_001073141.1 | ||||
NM_001278438.1 | Intron | NP_001265367.1 | ||||
NM_014923.4 | Intron | NP_055738.3 | ||||
XM_011534997.2 | Intron | XP_011533299.1 | ||||
XM_017020440.1 | Intron | XP_016875929.1 | ||||
XM_017020441.1 | Intron | XP_016875930.1 | ||||
XM_017020442.1 | Intron | XP_016875931.1 | ||||
XM_017020443.1 | Intron | XP_016875932.1 |
Set Membership: |
HapMap |