Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602965 MIM: 610728 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FABP7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FABP7 - fatty acid binding protein 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPDL3A - sphingomyelin phosphodiesterase acid like 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286138.1 | 782 | Intron | NP_001273067.1 | |||
NM_006714.4 | 782 | Intron | NP_006705.1 | |||
XM_005266797.4 | 782 | Intron | XP_005266854.1 | |||
XM_005266798.2 | 782 | Intron | XP_005266855.1 | |||
XM_005266799.2 | 782 | Intron | XP_005266856.1 | |||
XM_005266801.2 | 782 | Intron | XP_005266858.1 | |||
XM_005266802.2 | 782 | UTR 5 | XP_005266859.1 | |||
XM_017010203.1 | 782 | Intron | XP_016865692.1 | |||
XM_017010204.1 | 782 | Intron | XP_016865693.1 |
Set Membership: |
HapMap |