Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612677 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYHIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PYHIN1 - pyrin and HIN domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152501.4 | Intron | NP_689714.2 | ||||
NM_198928.4 | Intron | NP_945146.1 | ||||
NM_198929.4 | Intron | NP_945147.1 | ||||
NM_198930.3 | Intron | NP_945148.1 | ||||
XM_005244930.1 | Intron | XP_005244987.1 | ||||
XM_011509242.2 | Intron | XP_011507544.1 | ||||
XM_011509243.2 | Intron | XP_011507545.1 | ||||
XM_017000463.1 | Intron | XP_016855952.1 |
Set Membership: |
HapMap |