Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609449 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDE1 - nudE neurodevelopment protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143979.1 | Intron | NP_001137451.1 | ||||
NM_017668.2 | Intron | NP_060138.1 | ||||
XM_005255396.4 | Intron | XP_005255453.1 | ||||
XM_006720897.3 | Intron | XP_006720960.1 | ||||
XM_006720900.3 | Intron | XP_006720963.1 | ||||
XM_011522553.2 | Intron | XP_011520855.1 | ||||
XM_017023349.1 | Intron | XP_016878838.1 | ||||
XM_017023350.1 | Intron | XP_016878839.1 | ||||
XM_017023351.1 | Intron | XP_016878840.1 | ||||
XM_017023352.1 | Intron | XP_016878841.1 | ||||
XM_017023353.1 | Intron | XP_016878842.1 | ||||
XM_017023354.1 | Intron | XP_016878843.1 | ||||
XM_017023355.1 | Intron | XP_016878844.1 | ||||
XM_017023356.1 | Intron | XP_016878845.1 | ||||
XM_017023357.1 | Intron | XP_016878846.1 |