Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616073 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEPDC1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DEPDC1B - DEP domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145208.1 | 2096 | UTR 3 | NP_001138680.1 | |||
NM_018369.2 | 2096 | UTR 3 | NP_060839.2 | |||
XM_011543509.2 | 2096 | UTR 3 | XP_011541811.1 | |||
XM_011543510.2 | 2096 | Intron | XP_011541812.1 | |||
XM_017009629.1 | 2096 | Intron | XP_016865118.1 |
Set Membership: |
HapMap |