Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612935 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPRIP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MPRIP - myosin phosphatase Rho interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015134.3 | Intron | NP_055949.2 | ||||
NM_201274.3 | Intron | NP_958431.2 | ||||
XM_005256563.4 | Intron | XP_005256620.1 | ||||
XM_005256564.4 | Intron | XP_005256621.3 | ||||
XM_011523761.2 | Intron | XP_011522063.1 | ||||
XM_011523762.2 | Intron | XP_011522064.1 | ||||
XM_011523763.2 | Intron | XP_011522065.1 | ||||
XM_011523764.2 | Intron | XP_011522066.1 | ||||
XM_011523765.2 | Intron | XP_011522067.1 | ||||
XM_011523766.2 | Intron | XP_011522068.2 | ||||
XM_011523767.2 | Intron | XP_011522069.2 | ||||
XM_017024393.1 | Intron | XP_016879882.1 |
Set Membership: |
HapMap |