Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123930 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP2B6 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CYP2B6 - cytochrome P450 family 2 subfamily B member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000767.4 | Intron | NP_000758.1 | ||||
XM_005258569.4 | Intron | XP_005258626.1 | ||||
XM_006723050.3 | Intron | XP_006723113.1 | ||||
XM_011526546.2 | Intron | XP_011524848.1 | ||||
XM_011526547.2 | Intron | XP_011524849.1 | ||||
XM_011526548.2 | Intron | XP_011524850.1 | ||||
XM_011526549.2 | Intron | XP_011524851.1 | ||||
XM_011526550.2 | Intron | XP_011524852.1 |
Set Membership: |
HapMap |