Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEACAM21 - carcinoembryonic antigen related cell adhesion molecule 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098506.3 | Intron | NP_001091976.3 | ||||
NM_001288773.2 | Intron | NP_001275702.2 | ||||
NM_001290113.1 | Intron | NP_001277042.1 | ||||
NM_033543.5 | Intron | NP_291021.4 | ||||
XM_005278396.4 | Intron | XP_005278453.1 | ||||
XM_005278397.3 | Intron | XP_005278454.1 | ||||
XM_011527450.1 | Intron | XP_011525752.1 | ||||
XM_011527452.1 | Intron | XP_011525754.1 | ||||
XM_017027430.1 | Intron | XP_016882919.1 | ||||
XM_017027431.1 | Intron | XP_016882920.1 | ||||
XM_017027432.1 | Intron | XP_016882921.1 | ||||
XM_017027433.1 | Intron | XP_016882922.1 | ||||
XM_017027434.1 | Intron | XP_016882923.1 | ||||
XM_017027435.1 | Intron | XP_016882924.1 | ||||
XM_017027436.1 | Intron | XP_016882925.1 | ||||
XM_017027437.1 | Intron | XP_016882926.1 |
Set Membership: |
HapMap |