Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602320 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NELL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NELL2 - neural EGFL like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145107.1 | Intron | NP_001138579.1 | ||||
NM_001145108.1 | Intron | NP_001138580.1 | ||||
NM_001145109.1 | Intron | NP_001138581.1 | ||||
NM_001145110.1 | Intron | NP_001138582.1 | ||||
NM_006159.2 | Intron | NP_006150.1 | ||||
XM_005268905.3 | Intron | XP_005268962.1 | ||||
XM_011538396.1 | Intron | XP_011536698.1 | ||||
XM_017019341.1 | Intron | XP_016874830.1 | ||||
XM_017019342.1 | Intron | XP_016874831.1 | ||||
XM_017019343.1 | Intron | XP_016874832.1 | ||||
XM_017019344.1 | Intron | XP_016874833.1 |
Set Membership: |
HapMap |