Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190181 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFBR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGFBR1 - transforming growth factor beta receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130916.2 | Intron | NP_001124388.1 | ||||
NM_001306210.1 | Intron | NP_001293139.1 | ||||
NM_004612.3 | Intron | NP_004603.1 | ||||
XM_011518948.2 | Intron | XP_011517250.1 | ||||
XM_011518949.2 | Intron | XP_011517251.1 | ||||
XM_011518950.2 | Intron | XP_011517252.1 | ||||
XM_017015063.1 | Intron | XP_016870552.1 |