Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601752 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENTPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ENTPD1 - ectonucleoside triphosphate diphosphohydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098175.1 | Intron | NP_001091645.1 | ||||
NM_001164178.1 | Intron | NP_001157650.1 | ||||
NM_001164179.1 | Intron | NP_001157651.1 | ||||
NM_001164181.1 | Intron | NP_001157653.1 | ||||
NM_001164182.1 | Intron | NP_001157654.1 | ||||
NM_001164183.1 | Intron | NP_001157655.1 | ||||
NM_001312654.1 | Intron | NP_001299583.1 | ||||
NM_001320916.1 | Intron | NP_001307845.1 | ||||
NM_001776.5 | Intron | NP_001767.3 | ||||
XM_011540370.2 | Intron | XP_011538672.2 | ||||
XM_011540371.2 | Intron | XP_011538673.1 | ||||
XM_011540372.2 | Intron | XP_011538674.2 | ||||
XM_011540373.2 | Intron | XP_011538675.2 | ||||
XM_011540374.2 | Intron | XP_011538676.2 | ||||
XM_011540376.2 | Intron | XP_011538678.2 | ||||
XM_011540377.2 | Intron | XP_011538679.1 | ||||
XM_017016958.1 | Intron | XP_016872447.1 | ||||
XM_017016959.1 | Intron | XP_016872448.1 | ||||
XM_017016960.1 | Intron | XP_016872449.1 | ||||
XM_017016961.1 | Intron | XP_016872450.1 | ||||
XM_017016962.1 | Intron | XP_016872451.1 | ||||
XM_017016963.1 | Intron | XP_016872452.1 | ||||
XM_017016964.1 | Intron | XP_016872453.1 |
Set Membership: |
HapMap |