Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTAR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTAR1 - protein prenyltransferase alpha subunit repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099666.1 | 4410 | UTR 3 | NP_001093136.1 | |||
XM_005251976.4 | 4410 | UTR 3 | XP_005252033.1 | |||
XM_005251977.4 | 4410 | UTR 3 | XP_005252034.1 | |||
XM_005251980.4 | 4410 | Intron | XP_005252037.1 | |||
XM_005251981.4 | 4410 | Intron | XP_005252038.1 | |||
XM_011518639.1 | 4410 | UTR 3 | XP_011516941.1 | |||
XM_011518640.1 | 4410 | UTR 3 | XP_011516942.1 |
Set Membership: |
HapMap |