Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611352 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMBOX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HMBOX1 - homeobox containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135726.2 | Intron | NP_001129198.1 | ||||
NM_001324382.1 | Intron | NP_001311311.1 | ||||
NM_001324383.1 | Intron | NP_001311312.1 | ||||
NM_001324384.1 | Intron | NP_001311313.1 | ||||
NM_001324385.1 | Intron | NP_001311314.1 | ||||
NM_001324386.1 | Intron | NP_001311315.1 | ||||
NM_001324387.1 | Intron | NP_001311316.1 | ||||
NM_001324388.1 | Intron | NP_001311317.1 | ||||
NM_001324389.1 | Intron | NP_001311318.1 | ||||
NM_001324390.1 | Intron | NP_001311319.1 | ||||
NM_001324391.1 | Intron | NP_001311320.1 | ||||
NM_001324392.1 | Intron | NP_001311321.1 | ||||
NM_001324393.1 | Intron | NP_001311322.1 | ||||
NM_001324394.1 | Intron | NP_001311323.1 | ||||
NM_001324395.1 | Intron | NP_001311324.1 | ||||
NM_024567.4 | Intron | NP_078843.2 | ||||
XM_005273635.1 | Intron | XP_005273692.1 | ||||
XM_005273641.4 | Intron | XP_005273698.1 | ||||
XM_006716398.2 | Intron | XP_006716461.1 | ||||
XM_011544642.2 | Intron | XP_011542944.1 | ||||
XM_017013823.1 | Intron | XP_016869312.1 | ||||
XM_017013824.1 | Intron | XP_016869313.1 | ||||
XM_017013825.1 | Intron | XP_016869314.1 |
INTS9 - integrator complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |