Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611352 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INTS9 - integrator complex subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145159.2 | 2249 | UTR 3 | NP_001138631.1 | |||
NM_001172562.1 | 2249 | UTR 3 | NP_001166033.1 | |||
NM_018250.3 | 2249 | UTR 3 | NP_060720.2 | |||
XM_006716356.2 | 2249 | UTR 3 | XP_006716419.1 | |||
XM_006716357.3 | 2249 | Intron | XP_006716420.1 | |||
XM_011544573.1 | 2249 | UTR 3 | XP_011542875.1 | |||
XM_011544574.1 | 2249 | UTR 3 | XP_011542876.1 | |||
XM_011544575.2 | 2249 | UTR 3 | XP_011542877.1 | |||
XM_011544576.1 | 2249 | UTR 3 | XP_011542878.1 |