Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604913 MIM: 609927 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT7 - CCR4-NOT transcription complex subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS37A - VPS37A, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145152.1 | Intron | NP_001138624.1 | ||||
NM_152415.2 | Intron | NP_689628.2 | ||||
XM_005273400.2 | Intron | XP_005273457.1 | ||||
XM_005273401.2 | Intron | XP_005273458.1 | ||||
XM_006716286.1 | Intron | XP_006716349.1 | ||||
XM_017013021.1 | Intron | XP_016868510.1 | ||||
XM_017013022.1 | Intron | XP_016868511.1 | ||||
XM_017013023.1 | Intron | XP_016868512.1 | ||||
XM_017013024.1 | Intron | XP_016868513.1 | ||||
XM_017013025.1 | Intron | XP_016868514.1 | ||||
XM_017013026.1 | Intron | XP_016868515.1 |