Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607220 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTN6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNTN6 - contactin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289080.1 | Intron | NP_001276009.1 | ||||
NM_001289081.1 | Intron | NP_001276010.1 | ||||
NM_014461.3 | Intron | NP_055276.1 | ||||
XM_005265058.3 | Intron | XP_005265115.1 | ||||
XM_011533590.2 | Intron | XP_011531892.1 | ||||
XM_011533591.2 | Intron | XP_011531893.1 | ||||
XM_017006171.1 | Intron | XP_016861660.1 | ||||
XM_017006172.1 | Intron | XP_016861661.1 | ||||
XM_017006173.1 | Intron | XP_016861662.1 | ||||
XM_017006174.1 | Intron | XP_016861663.1 | ||||
XM_017006175.1 | Intron | XP_016861664.1 | ||||
XM_017006176.1 | Intron | XP_016861665.1 | ||||
XM_017006177.1 | Intron | XP_016861666.1 |
Set Membership: |
HapMap |