Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607220 | ||||||||||||||||||||
Literature Links: |
CNTN6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNTN6 - contactin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289080.1 | Intron | NP_001276009.1 | ||||
NM_001289081.1 | Intron | NP_001276010.1 | ||||
NM_014461.3 | Intron | NP_055276.1 | ||||
XM_005265058.3 | Intron | XP_005265115.1 | ||||
XM_011533590.2 | Intron | XP_011531892.1 | ||||
XM_011533591.2 | Intron | XP_011531893.1 | ||||
XM_017006171.1 | Intron | XP_016861660.1 | ||||
XM_017006172.1 | Intron | XP_016861661.1 | ||||
XM_017006173.1 | Intron | XP_016861662.1 | ||||
XM_017006174.1 | Intron | XP_016861663.1 | ||||
XM_017006175.1 | Intron | XP_016861664.1 | ||||
XM_017006176.1 | Intron | XP_016861665.1 | ||||
XM_017006177.1 | Intron | XP_016861666.1 |