Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613984 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FANCD2 - Fanconi anemia complementation group D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018115.2 | Intron | NP_001018125.1 | ||||
NM_001319984.1 | Intron | NP_001306913.1 | ||||
NM_033084.4 | Intron | NP_149075.2 |
FANCD2OS - FANCD2 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164839.1 | Intron | NP_001158311.1 | ||||
NM_173472.1 | Intron | NP_775743.1 | ||||
XM_011533330.2 | Intron | XP_011531632.1 | ||||
XM_011533331.2 | Intron | XP_011531633.1 |