Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606867 MIM: 611430 MIM: 612167 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GORASP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GORASP1 - golgi reassembly stacking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278789.1 | 2497 | UTR 3 | NP_001265718.1 | |||
NM_001278790.1 | 2497 | UTR 3 | NP_001265719.1 | |||
NM_031899.3 | 2497 | UTR 3 | NP_114105.1 | |||
XM_006713301.3 | 2497 | UTR 3 | XP_006713364.1 | |||
XM_011534020.2 | 2497 | UTR 3 | XP_011532322.1 | |||
XM_011534021.2 | 2497 | UTR 3 | XP_011532323.1 | |||
XM_017007050.1 | 2497 | UTR 3 | XP_016862539.1 | |||
XM_017007051.1 | 2497 | UTR 3 | XP_016862540.1 | |||
XM_017007052.1 | 2497 | UTR 3 | XP_016862541.1 |
TTC21A - tetratricopeptide repeat domain 21A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR48 - WD repeat domain 48 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |