Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609938 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CADM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CADM2 - cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167674.1 | Intron | NP_001161146.1 | ||||
NM_001167675.1 | Intron | NP_001161147.1 | ||||
NM_001256502.1 | Intron | NP_001243431.1 | ||||
NM_001256503.1 | Intron | NP_001243432.1 | ||||
NM_001256504.1 | Intron | NP_001243433.1 | ||||
NM_001256505.1 | Intron | NP_001243434.1 | ||||
NM_153184.3 | Intron | NP_694854.2 | ||||
XM_006713081.3 | Intron | XP_006713144.1 | ||||
XM_006713082.3 | Intron | XP_006713145.1 | ||||
XM_017006062.1 | Intron | XP_016861551.1 | ||||
XM_017006063.1 | Intron | XP_016861552.1 |