Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600526 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MARK2 - microtubule affinity regulating kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039469.2 | Intron | NP_001034558.2 | ||||
NM_001163296.1 | Intron | NP_001156768.1 | ||||
NM_001163297.1 | Intron | NP_001156769.1 | ||||
NM_004954.4 | Intron | NP_004945.4 | ||||
NM_017490.3 | Intron | NP_059672.2 | ||||
XM_011544789.1 | Intron | XP_011543091.1 | ||||
XM_011544790.1 | Intron | XP_011543092.1 | ||||
XM_011544791.1 | Intron | XP_011543093.1 | ||||
XM_011544792.1 | Intron | XP_011543094.1 | ||||
XM_011544793.1 | Intron | XP_011543095.1 | ||||
XM_011544794.1 | Intron | XP_011543096.1 | ||||
XM_011544795.1 | Intron | XP_011543097.1 | ||||
XM_011544796.1 | Intron | XP_011543098.1 | ||||
XM_011544797.1 | Intron | XP_011543099.1 | ||||
XM_011544798.1 | Intron | XP_011543100.1 | ||||
XM_017017310.1 | Intron | XP_016872799.1 | ||||
XM_017017311.1 | Intron | XP_016872800.1 |
Set Membership: |
HapMap |