Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610593 MIM: 607858 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP6D1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP6D1 - MAP6 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARL - presenilin associated rhomboid like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037639.2 | Intron | NP_001032728.1 | ||||
NM_001324436.1 | Intron | NP_001311365.1 | ||||
NM_001324437.1 | Intron | NP_001311366.1 | ||||
NM_001324438.1 | Intron | NP_001311367.1 | ||||
NM_018622.6 | Intron | NP_061092.3 | ||||
XM_005247582.4 | Intron | XP_005247639.1 | ||||
XM_017006800.1 | Intron | XP_016862289.1 | ||||
XM_017006801.1 | Intron | XP_016862290.1 | ||||
XM_017006802.1 | Intron | XP_016862291.1 | ||||
XM_017006803.1 | Intron | XP_016862292.1 |
Set Membership: |
HapMap |